Läs mer
Thiorphan, TRC
Produktkod. 30132613
Change view
Kvantitet:
5 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30132613 25 mg
30135671 100 mg
30006588 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30132613 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT36950025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 76721-89-6
Molekylformel C12 H15 N O3 S
Molekylvikt (g/mol) 253.32
Synonym Glycine, N-[2-(mercaptomethyl)-1-oxo-3-phenylpropyl]-, (±)-, (±)-Thiorphan, DL-Thiorphan, N-[2-(Mercaptomethyl)-1-oxo-3-phenylpropyl]glycine, Thiorphan
IUPAC-namn 2-[(2-benzyl-3-sulfanylpropanoyl)amino]acetic acid
LEDER OC(=O)CNC(=O)C(CS)Cc1ccccc1
Specifikationer
Kemiskt namn eller material Thiorphan
Anteckningar om renhetsgrad HPLC
InChI formel InChI=1S/C12H15NO3S/c14-11(15)7-13-12(16)10(8-17)6-9-4-2-1-3-5-9/h1-5,10,17H,6-8H2,(H,13,16)(H,14,15)
Kvantitet 25 mg
Formel vikt 253.0773
Procent renhet >95%