Läs mer
Thiosildenafil, LoGiCal
Produktkod. 30462367
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30462367 10 mg
1 options
Produktkod. 30462367 Leverantör LoGiCal Leverantörsnummer LGCFOR0254.39
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Thiosildenafil
CAS 479073-79-5
Analyt- eller komponentnamn Thiosildenafil
Frakt skick Room Temperature
Molekylformel C22 H30 N6 O3 S2
Certifikat och efterlevnad ISO 9001
Synonym 7H-Pyrazolo[4,3-d]pyrimidine-7-thione, 5-[2-ethoxy-5-[(4-methyl-1-piperazinyl)sulfonyl]phenyl]-1,6-dihydro-1-methyl-3-propyl-, Piperazine, 1-[[3-(4,7-dihydro-1-methyl-3-propyl-7-thioxo-1H-pyrazolo[4,3-d]pyrimidin-5-yl)-4-ethoxyphenyl]sulfonyl]-4-methyl- (9CI), Thiosildenafil
InChI formel InChI=1S/C22H30N6O3S2/c1-5-7-17-19-20(27(4)25-17)22(32)24-21(23-19)16-14-15(8-9-18(16)31-6-2)33(29,30)28-12-10-26(3)11-13-28/h8-9,14H,5-7,10-13H2,1-4H3,(H,23,24,32)
Lösningstyp Neat
LEDER CCCc1nn(C)c2C(=S)NC(=Nc12)c3cc(ccc3OCC)S(=O)(=O)N4CCN(C)CC4
IUPAC-namn 5-[2-ethoxy-5-(4-methylpiperazin-1-yl)sulfonylphenyl]-1-methyl-3-propyl-6H-pyrazolo[4,3-d]pyrimidine-7-thione
Industry Type Clinical Testing
Molekylvikt (g/mol) 490.64
Kvantitet 10 mg
Formel vikt 490.1821 g/mol