Läs mer
Thiouric Acid-13C3 Sodium Salt Dihydrate, TRC
Produktkod. 30453556
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30453556 10 mg
30454105 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30453556 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT37550210MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1330823-16-9
Molekylformel 13C3 C2 H3 N4 Na O2 S
Molekylvikt (g/mol) 209.136
IUPAC-namn (2,8-dihydroxy-9H-purin-6-yl)sulfanylsodium
LEDER O[13c]1n[13c]2[13c](S[Na])nc(O)nc2[nH]1
Specifikationer
Kemiskt namn eller material Thiouric Acid-13C3 Sodium Salt Dihydrate
Kvantitet 10 mg
InChI formel InChI=1S/C5H4N4O2S.Na/c10-4-6-1-2(7-4)8-5(11)9-3(1)12;/h(H4,6,7,8,9,10,11,12);/q;+1/p-1/i1+1,3+1,4+1;
Formel vikt 208.998