Läs mer
Thiouric Acid Sodium Salt Dihydrate, TRC
Produktkod. 30449248
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30449248 100 mg
1 options
Produktkod. 30449248 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT375500100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Thiouric Acid Sodium Salt Dihydrate
CAS 1329805-85-7
Anteckningar om renhetsgrad HPLC
Molekylformel C5 H3 N4 O2 S . Na . 2 H2 O
Kvantitet 100 mg
Synonym 7,9-Dihydro-6-thioxo-1H-purine-2,8(3H,6H)-dione Sodium Salt Dihydrate, 6-Thio-uric Acid Sodium Dihydrate, 2,8-Dihydroxy-6-mercaptopurine Sodium Dihydrate, 6-Thiouric Acid Sodium Dihydrate, NSC 46380
InChI formel InChI=1S/C5H4N4O2S.Na.2H2O/c10-4-6-1-2(7-4)8-5(11)9-3(1)12;;;/h(H4,6,7,8,9,10,11,12);;2*1H2/q;+1;;/p-1
LEDER O.O.[Na+].Oc1nc([S-])c2nc(O)[nH]c2n1
IUPAC-namn sodium;2,8-dihydroxy-9H-purine-6-thiolate;dihydrate
Molekylvikt (g/mol) 242.19
Formel vikt 242.0086