Learn More
Invitrogen™ THRA Human Protein, Ligand Binding Domain, GST-Tagged
Thyroid Receptor alpha, Ligand Binding Domain (THRA LBD ) is a 57.90 kDa recombinant human protein (amino acids 148-410) expressed as a GST-tagged fusion protein in baculovirus-infected insect cells.
Brand:  Invitrogen™ PV4762
Additional Details : Weight : 0.50000kg
Product Code. 10702344
  • 17690.00 SEK / Each
Estimated Shipment: 13-05-2024
to see stock.
For Research Use Only. All usage must comply with product instructions.
Description
Description
THRA LBD was expressed and purified in the absence of exogenous ligands, making it ideal for ligand binding and coregulator displacement studies.
Specifications
Specifications
Nuclear Receptor Assay
Store in ultra-cold freezer (-68 to -85°C).
Ligand Binding Domain
Human
Hormone Receptors
50 μg
RUO - research use only
TR-α LBD
Recombinant
Product Suggestions