Learn More
Buerkle™ Thread Adapter for Mini Solvent Pump
Thread adapter for Buerkle™ Mini solvent pump.
Brand: Buerkle™ 0006-4003
508.74 SEK valid until 2024-12-31
Use promo code "23267" to get your promotional price.
Additional Details : Weight : 0.02500kg
Description
- For use with Buerkle Mini Solvent Pump
- ETFE Construction
- External thread: GL 45
Specifications
Mini Solvent Pump | |
ETFE | |
GL45-GL32 | |
Thread Adapter |