Learn More
SCAT™ Polypropylene Thread Adapters
Safety Caps and Safety Waste Caps are available in many thread sizes. If the right thread for your container is still not available, simply use the matching adapter.
Brand: SCAT™ 107028
722.00 SEK valid until 2024-05-31
Use promo code "21679" to get your promotional price.
Additional Details : Weight : 0.01000kg
Description
SCAT™ thread adapters are ideal for connecting safety caps, safety waste caps, funnels, canisters or other disposal and supply units.
- Made of polypropylene (PP) material
- Available in a wide range of thread sizes
- Suitable for all SCAT™ caps, funnels or canisters
Specifications
Thread 1: R 1 1/2 in. (f), Thread 2: GL45 (m) | |
Colorless | |
Polypropylene | |
Thread Adapter |