Learn More
Lenz Laborglasintrumente™ Three-Neck Round-Bottom Flasks
Three-neck round-bottom reaction flasks with two parallel side necks
563.92 SEK - 9969.30 SEK
Specifications
Material | Borosilicate Glass (3.3) |
---|---|
No. per Pack | 1 |
Product Type | Reaction flasks |
Product Code | Brand | Capacity (Metric) | Description | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Description | Price | Quantity & Availability | ||||
10121761
|
Lenz Laborglasintrumente™
03304237 |
100 mL | Center neck- NS 29/32 , Side necks -NS 19/26 |
563.92 SEK
Each |
Estimated Shipment: 11-06-2024
Log in to see stock. |
||||
10264191
|
Lenz Laborglasintrumente™
03304170 |
1000 mL | Center neck- NS 29/32, Side necks - NS 14/23 |
869.20 SEK
Each |
Estimated Shipment: 11-06-2024
Log in to see stock. |
||||
10709031
|
Lenz Laborglasintrumente™
03305476 |
3000 mL | Center neck- NS 45/40 , Side necks -NS 29/32 |
2538.70 SEK
Each |
Estimated Shipment: 22-05-2024
Log in to see stock. |
||||
10718841
|
Lenz Laborglasintrumente™
03304479 |
4000 mL | Center neck- NS 29/32, Side necks - NS 29/32 |
2559.90 SEK
Each |
Estimated Shipment: 28-05-2024
Log in to see stock. |
||||
10798451
|
Lenz Laborglasintrumente™
03305494 |
20,000 mL | Center neck- NS 45/40 , Side necks -NS 29/32 |
9969.30 SEK
Each |
Estimated Shipment: 11-06-2024
Log in to see stock. |
||||
Description
- Reaction flasks acc. to DIN 12392 and in addition to DIN
- With two parallel side necks
- With conical ground joints
- With inscription label
- Material: DURAN™
Specifications
Borosilicate Glass (3.3) | |
1 | |
Reaction flasks |