missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thrombin Bovine High Purity Grade MP Biomedicals
Click to view available options
Quantity:
1 kU
Specifications
CAS | 9002-12-4 |
Assay Percent Range | 10% |
Molecular Formula | C18H26N5O14P |
Quantity | 1 kU |
Synonym | uricase, urate oxidase, urate:o2 oxidoreductase, uricase, candida utilis, uricase from candida sp., urate: oxygen oxoreductase, uricase from bacillus fastidiosus, uricase from arthrobacter globiformis, upu, uricase from bacillus fastidiosus, lyophilized, ∼9 u/mg |
InChI Key | VPXDHZMLJOJKOX-UHFFFAOYNA-N |
SMILES | C1=CN(C(=O)NC1=O)C2C(C(C(O2)COP(=O)(O)OC3C(OC(C3O)N4C=CC(=O)NC4=O)CO)O)O.N |
IUPAC Name | azane;[5-(2,4-dioxopyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl [5-(2,4-dioxopyrimidin-1-yl)-4-hydroxy-2-(hydroxymethyl)oxolan-3-yl] hydrogen phosphate |
Molecular Weight (g/mol) | 567.40 |
PubChem CID | 14228836 |
Show More |
Safety and Handling
Product Identifier
- Thrombin, Bovine, High Purity Grade
- Danger
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H335-May cause respiratory irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : thrombin
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'