Läs mer
Thymidine-1',2',3',4',5'-13C5, TRC
Produktkod. 30454486
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30454486 1 mg
30454391 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30454486 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT4119971MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 156968-81-9
Molekylformel 13C5 C5 H14 N2 O5
Molekylvikt (g/mol) 247.19
Synonym 1-(2-Deoxy-β-D-erythro-pentofuranosyl)-5-methyl-2,4(1H,3H)-pyrimidinedione-1',2',3',4',5'-13C5, 2'-Deoxythymidine-3'-13C, 5-Methyl-2'-deoxyuridine-1',2',3',4',5'-13C5, 5-Methyldeoxyuridine-1',2',3',4',5'-13C5, DThyd-1',2',3',4',5'-13C5, Deoxyribothymidine-1',2',3',4',5'-13C5, Deoxythymidine-1',2',3',4',5'-13C5, NSC 21548-1',2',3',4',5'-13C5, Thymidin-1',2',3',4',5'-13C5, Thymine 2-Desoxyriboside-1',2',3',4',5'-13C5, Thymine Deoxyriboside-1',2',3',4',5'-13C5, 2'-Deoxy-5-methyl-uridine-1',2',3',4',5'-13C5, dT-1',2',3',4',5'-13C5, Thymine-1,2-deoxy-β-D-ribofuranoside-1',2',3',4',5'-13C5
IUPAC-namn 1-[(2R,4S,5R)-4-hydroxy-5-(hydroxy(1^{13}C)methyl)(2,3,4,5-^{13}C_{4})oxolan-2-yl]-5-methylpyrimidine-2,4-dione
LEDER CC1=CN([13C@H]2[13CH2][13C@H](O)[13C@@H]([13CH2]O)O2)C(=O)NC1=O
Specifikationer
Kemiskt namn eller material Thymidine-1',2',3',4',5'-13C5
Kvantitet 1 mg
InChI formel InChI=1S/C10H14N2O5/c1-5-3-12(10(16)11-9(5)15)8-2-6(14)7(4-13)17-8/h3,6-8,13-14H,2,4H2,1H3,(H,11,15,16)/t6-,7+,8+/m0/s1/i2+1,4+1,6+1,7+1,8+1
Formel vikt 247.107