Läs mer
Thymine-2-13C-1,3-15N2, TRC
Produktkod. 30453456
Change view
Kvantitet:
2.5 mg
50 mg
100 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30453456 100 mg -
30452980 50 mg -
30451345 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30453456 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT412149100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1397236-31-5
Molekylformel C4 13C H6 15N2 O2
Molekylvikt (g/mol) 129.09
Synonym 5-Methyl-2,4(1H,3H)-pyrimidinedione-2-13C-1,3-15N2
LEDER O=C1C(C)=C[15N]([H])[13C]([15N]1[H])=O
Specifikationer
Kemiskt namn eller material Thymine-2-13C-1,3-15N2
Kvantitet 100 mg
InChI formel InChI=1S/C5H6N2O2/c1-3-2-6-5(9)7-4(3)8/h2H,1H3,(H2,6,7,8,9)/i5+1,6+1,7+1
Formel vikt 129.0404