Läs mer
Thymol-b-D-glucopyranoside, TRC
Produktkod. 30451018
Change view
Kvantitet:
25 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30451018 25 mg
30450253 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30451018 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT41309025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Thymol-b-D-glucopyranoside
CAS 20772-23-0
Molekylformel C16H24O6
Kvantitet 25 mg
InChI formel InChI=1S/C16H24O6/c1-8(2)10-5-4-9(3)6-11(10)21-16-15(20)14(19)13(18)12(7-17)22-16/h4-6,8,12-20H,7H2,1-3H3/t12-,13-,14+,15-,16-/m1/s1
LEDER CC(C)c1ccc(C)cc1O[C@@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O
IUPAC-namn (2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-(5-methyl-2-propan-2-ylphenoxy)oxane-3,4,5-triol
Molekylvikt (g/mol) 312.36
Formel vikt 312.1573