missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thymol Blue Indicator, Honeywell Fluka™
668.00 SEK valid until 2024-04-26
Use promo code "21687" to get your promotional price.
Use promo code "21687" to get your promotional price.
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass Bottle
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Melting Point | 221°C to 224°C |
UN Number | NONH for all modes of transport |
Beilstein | 368036 |
Formula Weight | 466.59g/mol |
Packaging | Glass Bottle |
Visual Transition Interval | 1.2 to 2.8 (red to yellow) |
Density | 0.979g/cm3 |
Quantity | 5 g |
Physical Form | Powder |
Chemical Name or Material | Thymol Blue |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'