Läs mer
Thymol O-(beta)-D-Glucuronide, TRC
Produktkod. 30453949
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30453949 10 mg
1 options
Produktkod. 30453949 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT41305010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Thymol O-b-D-Glucuronide
CAS 7702-71-8
Molekylformel C16H22O7
Kvantitet 10 mg
Synonym 5-Methyl-2-(1-methylethyl)phenyl β-D-Glucopyranosiduronic Acid
InChI formel InChI=1S/C16H22O7/c1-7(2)9-5-4-8(3)6-10(9)22-16-13(19)11(17)12(18)14(23-16)15(20)21/h4-7,11-14,16-19H,1-3H3,(H,20,21)/t11-,12-,13+,14-,16+/m0/s1
LEDER O[C@@H]([C@H](O)[C@H]1O)[C@@H](C(O)=O)O[C@H]1OC2=C(C(C)C)C=CC(C)=C2
IUPAC-namn (2S,3S,4S,5R,6S)-3,4,5-trihydroxy-6-(2-isopropyl-5-methylphenoxy)tetrahydro-2H-pyran-2-carboxylic acid
Molekylvikt (g/mol) 326.34
Formel vikt 326.14