Learn More
Thermo Scientific™ Thymopoietin II Peptide Fragment
For cellular and molecular biology applications
Marke: Thermo Scientific™ J66498.|D
Weitere Details : CAS Number : 56795-64-3 Weight : 0.00001kg
Beschreibung
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Spezifikation
HPLC | |
Solid | |
Thymopoietin II Peptide Fragment | |
RUO - research use only | |
Gly-Glu-Gln-Arg-Lys-Asp-Val-Tyr-Val-Gln-Leu-Tyr-Leu |
-30°C to -10°C | |
1610.83 | |
0.5 mg | |
Light sensitive |
For Research Use Only