Läs mer
Thyroid Peroxidase Antibody (TPO28), Alexa Fluor™ 647, Novus Biologicals™
Handla allt Bio Techne ProdukterBeskrivning
Thyroid Peroxidase Monoclonal specifically detects Thyroid Peroxidase in Human samples. It is validated for Western Blot, ELISA, Sandwich ELISA.
Specifikationer
Specifikationer
| Antigen | Thyroid Peroxidase |
| Användningsområden | Western Blot, ELISA |
| Klassificering | Monoclonal |
| Klona | TPO28 |
| Konjugera | Alexa Fluor 647 |
| Utspädning | Western Blot, ELISA, Sandwich ELISA |
| Formulering | 50mM Sodium Borate with 0.05% Sodium Azide |
| Gene Alias | EC 1.11.1, MSA, TDH2A, thyroid microsomal antigen, thyroid peroxidase, thyroperoxidase, TPXEC 1.11.1.8 |
| Gensymboler | TPO |
| Värd art | Mouse |
| Visa mer |