Läs mer
Thyroxine Sodium Salt, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Thyroxine sodium salt |
| CAS | 55-03-8 |
| Molekylformel | C15 H10 I4 N O4 . Na |
| Kvantitet | 25 g |
| Synonym | L-Tyrosine','O-(4-hydroxy-3,5-diiodophenyl)-3,5-diiodo-','sodium salt (1:1), L-Tyrosine','O-(4-hydroxy-3,5-diiodophenyl)-3,5-diiodo-','monosodium salt (9CI), Thyroxine','monosodium salt','L- (8CI), Berlthyrox, Dathroid, Droxine, Eferox, Elthyrone, Elthyroxine, Eltroxin, Euthyrox, Eutirox, L-Thyroxin Henning, L-Thyroxine monosodium salt, L-Thyroxine sodium, L-Thyroxine sodium salt, Laevoxin, Letrox, Letter, Levaxin, Leventa, Levo-T, Levolet, Levoroxine, Levothroid, Levothyrox, Levothyroxine sodium, Levotirox, Levotiroxina, Levoxyl, Monosodium thyroxine, NSC 259940, Novothyrox, Oroxine, Puran T 4, Sodium L-thyroxine, Sodium levothyroxine, Sodium thyroxin, Sodium thyroxinate, Sodium thyroxine, Synthroid, Synthroid sodium, Synthrox, T 4KP, T4 Montpellier 100, Thevier, Throxinique, Thyradin, Thyradin S, Thyrax Duotab, Thyrex, Thyro 4, Thyrosit, Thyroxevan, Thyroxine sodium, Thyroxine sodium salt, Tiroidine, Tirosint, Unithroid, Levothyroxine Sodium Salt |
| InChI formel | InChI=1S/C15H11I4NO4.Na/c16-8-4-7(5-9(17)13(8)21)24-14-10(18)1-6(2-11(14)19)3-12(20)15(22)23;/h1-2,4-5,12,21H,3,20H2,(H,22,23);/q;+1/p-1/t12-;/m0./s1 |
| LEDER | [Na+].N[C@@H](Cc1cc(I)c(Oc2cc(I)c(O)c(I)c2)c(I)c1)C(=O)[O-] |
| IUPAC-namn | sodium;(2S)-2-amino-3-[4-(4-hydroxy-3,5-diiodophenoxy)-3,5-diiodophenyl]propanoate |
| Molekylvikt (g/mol) | 798.85 |
| Formel vikt | 798.6686 |