Läs mer
THZ1, MedChemExpress
Produktkod. 16497966
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
50 mg
100 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16497966 100 mg
16448248 10 mg
16430698 5 mg
16424137 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Produktkod. 16497966 Leverantör Medchem Express Leverantörsnummer HY80013
MedChemExpress THZ1 is a selective and potent covalent CDK7 inhibitor with an IC50 of 3.2 nM. THZ1 also inhibits the closely related kinases CDK12 and CDK13 and downregulates MYC expression.
THZ1 is a selective and potent covalent CDK7 inhibitor with an IC50 of 3.2 nM. THZ1 also inhibits the closely related kinases CDK12 and CDK13 and downregulates MYC expression.
Specifikationer
Kemiskt namn eller material THZ1
Fysisk form Solid
CAS 1604810-83-4
Färg Off-White
Kvantitet 100 mg
Anteckningar om renhetsgrad Research
Molekylformel C31H28ClN7O2
För användning med (applikation) Cancer-Kinase/protease
Löslighetsinformation DMSO : 100 mg/mL (176.66 mM; Need ultrasonic) ∣Ethanol : < 1 mg/mL (insoluble)
LEDER ClC1=CN=C(NC2=CC(NC(C3=CC=C(NC(/C=C/CN(C)C)=O)C=C3)=O)=CC=C2)N=C1C4=CNC5=CC=CC=C54
Molekylvikt (g/mol) 566.05
Formel vikt 566.05
Procent renhet 95.58%