Läs mer
THZ531, MedChemExpress
Produktkod. 16456597
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16456597 5 mg
16422886 25 mg
16403878 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 16456597 Leverantör Medchem Express Leverantörsnummer HY103618
MedChemExpress THZ531 is a selective and covalent inhibitor of both CDK12 and CDK13 with IC50s of 158 nM and 69 nM, respectively.
THZ531 is a selective and covalent inhibitor of both CDK12 and CDK13 with IC50s of 158 nM and 69 nM, respectively.
Specifikationer
Kemiskt namn eller material THZ531
CAS 1702809-17-3
Färg Off-White
Anteckningar om renhetsgrad Research
Molekylformel C30H32ClN7O2
Kvantitet 5 mg
Löslighetsinformation DMSO : 14.29 mg/mL (25.61 mM; Need ultrasonic)
LEDER ClC(C(C1=CNC2=CC=CC=C12)=N3)=CN=C3N[C@@H]4CCCN(C(C5=CC=C(NC(/C=C/CN(C)C)=O)C=C5)=O)C4
Molekylvikt (g/mol) 558.07
Formel vikt 558.07
Procent renhet 98.11%
Kvalitet Research
Fysisk form Powder
För användning med (applikation) Cancer-Kinase/protease