Läs mer
Tiamulin Fumarate, TRC
Produktkod. 30456596
Change view
Kvantitet:
2.5 g
Produktkod. Kvantitet
30456596 2.5 g
1 options
Produktkod. 30456596 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT4366002.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Tiamulin fumarate
CAS 55297-96-6
Anteckningar om renhetsgrad HPLC
Molekylformel C28 H47 N O4 S . C4 H4 O4
Kvantitet 2.5 g
Synonym Tiamulin Hydrogen Fumarate, [[2-(Diethylamino)ethyl]thio]acetic acid (3aS,4R,5S,6S,8R,9R,9aR,10R)-6-ethenyldecahydro-5-hydroxy-4,6,9,10-tetramethyl-1-oxo-3a,9-propano-3aH-cyclopentacycloocten-8-yl ester (2E)-2-butenedioate (1:1) (salt), [[2-(Diethylamino)ethyl]thio]acetic acid [3aS-(3aα,4β,5α,6α,8β,9α,9aβ,10S*)]-6-ethenyldecahydro-5-hydroxy-4,6,9,10-tetramethyl-1-oxo-3a,9-propano-3aH-cyclopentacycloocten-8-yl ester (E)-2-butenedioate (1:1) (salt), 3a,9-Propano-3aH-cyclopentacyclooctene','acetic acid deriv., 14-Deoxy-14-[(2-diethylaminoethyl)-mercaptoacetoxy]mutilin hydrogen fumarate, 81723hfu, Dynamutilin, SQ 22947, Sandoz 81723hfu, Thiamutilin fumarate, Tiamulin fumarate, Tiamulin hydrogen fumarate, Tiamutin, Tiavet, Vievitin
InChI formel InChI=1S/C28H47NO4S.C4H4O4/c1-8-26(6)17-22(33-23(31)18-34-16-15-29(9-2)10-3)27(7)19(4)11-13-28(20(5)25(26)32)14-12-21(30)24(27)28;5-3(6)1-2-4(7)8/h8,19-20,22,24-25,32H,1,9-18H2,2-7H3;1-2H,(H,5,6)(H,7,8)/b;2-1+/t19-,20+,22-,24+,25+,26-,27+,28+;/m1./s1
LEDER CCN(CC)CCSCC(=O)O[C@@H]1C[C@@](C)(C=C)[C@@H](O)[C@H](C)[C@@]23CC[C@@H](C)[C@]1(C)[C@@H]2C(=O)CC3.OC(=O)\C=C\C(=O)O
IUPAC-namn (3aS,4R,5S,6S,8R,9R,9aR,10R)-6-Ethenyl-5-hydroxy- 4,6,9,10-tetramethyl-1-oxodecahydro-3a,9-propano-3aH-cyclopentacycloocten-8-yl [[2-(diethylamino)ethyl]sulfanyl]acetate monofumarate
Molekylvikt (g/mol) 609.81
Formel vikt 609.3335