Läs mer
Tiamulin, TRC
Produktkod. 30006500
Change view
Kvantitet:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30006500 1 g
17981515 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30006500 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT4365951G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 55297-95-5
Molekylformel C28 H47 N O4 S
Molekylvikt (g/mol) 493.74
Synonym Tiamulin, (3aS,4R,5S,6S,8R,9R,9aR,10R)-6-Ethenyl-5-hydroxy- 4,6,9,10-tetramethyl-1-oxodecahydro-3a,9-propano-3aH-cyclopentacycloocten-8-yl [[2-(diethylamino)ethyl]sulfanyl]acetate, Acetic acid, 2-[[2-(diethylamino)ethyl]thio]-, (3aS,4R,5S,6S,8R,9R,9aR,10R)-6-ethenyldecahydro-5-hydroxy-4,6,9,10-tetramethyl-1-oxo-3a,9-propano-3aH-cyclopentacycloocten-8-yl ester, Acetic acid, [[2-(diethylamino)ethyl]thio]-, (3aS,4R,5S,6S,8R,9R,9aR,10R)-6-ethenyldecahydro-5-hydroxy-4,6,9,10-tetramethyl-1-oxo-3a,9-propano-3aH-cyclopentacycloocten-8-yl ester (9CI), Acetic acid, [[2-(diethylamino)ethyl]thio]-, 6-ethenyldecahydro-5-hydroxy-4,6,9,10-tetramethyl-1-oxo-3a,9-propano-3aH-cyclopentacycloocten-8-yl ester, [3aS-(3aα,4β,5α,6α,8β,9α,9aβ,10S*)]-, 3a,9-Propano-3aH-cyclopentacyclooctene, acetic acid deriv., Denagard, Thiamutilin, Tiavet P
IUPAC-namn (3aS,4R,5S,6S,8R,9R,9aR,10R)-6-Ethenyl-5-hydroxy- 4,6,9,10-tetramethyl-1-oxodecahydro-3a,9-propano-3aH-cyclopentacycloocten-8-yl [[2-(diethylamino)ethyl]sulfanyl]acetate
LEDER CCN(CC)CCSCC(=O)O[C@@H]1C[C@@](C)(C=C)[C@@H](O)[C@H](C)[C@@]23CC[C@@H](C)[C@]1(C)[C@@H]2C(=O)CC3
Specifikationer
Kemiskt namn eller material Tiamulin
Anteckningar om renhetsgrad HPLC
Kvantitet 1 g
InChI formel InChI=1S/C28H47NO4S/c1-8-26(6)17-22(33-23(31)18-34-16-15-29(9-2)10-3)27(7)19(4)11-13-28(20(5)25(26)32)14-12-21(30)24(27)28/h8,19-20,22,24-25,32H,1,9-18H2,2-7H3/t19-,20+,22-,24+,25+,26-,27+,28+/m1/s1
Formel vikt 493.3226
Procent renhet >95%