Läs mer
Ticagrelor Amine Derivative, TRC
Produktkod. 30451562
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30451562 100 mg
1 options
Produktkod. 30451562 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT437750100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Ticagrelor Amine Derivative
CAS 1902184-02-4
Molekylformel C23H29F2N7O3S
Kvantitet 100 mg
Synonym (1S,2S,3S,5R)-3-(2-Aminoethoxy)-5-(7-(((1R,2S)-2-(3,4-difluorophenyl)cyclopropyl)amino)-5-(propylthio)-3H-[1,2,3]triazolo[4,5-d]pyrimidin-3-yl)cyclopentane-1,2-diol
InChI formel InChI=1S/C23H29F2N7O3S/c1-2-7-36-23-28-21(27-15-9-12(15)11-3-4-13(24)14(25)8-11)18-22(29-23)32(31-30-18)16-10-17(35-6-5-26)20(34)19(16)33/h3-4,8,12,15-17,19-20,33-34H,2,5-7,9-10,26H2,1H3,(H,27,28,29)/t12-,15+,16+,17-,19-,20+/m0/s1
LEDER CCCSc1nc(N[C@@H]2C[C@H]2c3ccc(F)c(F)c3)c4nnn([C@@H]5C[C@H](OCCN)[C@@H](O)[C@H]5O)c4n1
IUPAC-namn (1S,2S,3S,5R)-3-(2-aminoethoxy)-5-[7-[[(1R,2S)-2-(3,4-difluorophenyl)cyclopropyl]amino]-5-propylsulfanyltriazolo[4,5-d]pyrimidin-3-yl]cyclopentane-1,2-diol
Molekylvikt (g/mol) 521.58
Formel vikt 521.2021