Läs mer
Ticlopidine EP Impurity G (as Hydrochloride), Mikromol™
Beskrivning
Ticlopidine EP Impurity G (as Hydrochloride)
Specifikationer
Specifikationer
| Kemiskt namn eller material | 5-(3-Chlorobenzyl)-4,5,6,7-tetrahydrothieno[3,2-c]pyridine Hydrochloride |
| CAS | 54943-16-7 |
| Analyt- eller komponentnamn | 5-(3-Chlorobenzyl)-4,5,6,7-tetrahydrothieno[3,2-c]pyridine Hydrochloride |
| Frakt skick | Room Temperature |
| Molekylformel | C14 H14 Cl N S . Cl H |
| Certifikat och efterlevnad | ISO 9001 |
| Synonym | Ticlopidine Hydrochloride Imp. G (EP), Ticlopidine Imp. G (EP), 5-(3-Chlorobenzyl)-4,5,6,7-tetrahydrothieno[3,2-c]pyridine Hydrochloride, Ticlopidine Hydrochloride Imp. G (EP) as Hydrochloride, Ticlopidine Hydrochloride Impurity G as Hydrochloride, Ticlopidine Impurity G as Hydrochloride |
| InChI formel | InChI=1S/C14H14ClNS.ClH/c15-13-3-1-2-11(8-13)9-16-6-4-14-12(10-16)5-7-17-14;/h1-3,5,7-8H,4,6,9-10H2;1H |
| LEDER | Cl.Clc1cccc(CN2CCc3sccc3C2)c1 |
| IUPAC-namn | 5-[(3-chlorophenyl)methyl]-6,7-dihydro-4H-thieno[3,2-c]pyridine;hydrochloride |
| Visa mer |