Läs mer
Ticlopidine EP Impurity J (as Dihydrochloride), Mikromol™
Beskrivning
Ticlopidine EP Impurity J (as Dihydrochloride)
Specifikationer
Specifikationer
| Kemiskt namn eller material | N,N'-Bis(2-chlorobenzyl)ethane-1,2-diamine Dihydrochloride |
| CAS | 2056-18-0 |
| Analyt- eller komponentnamn | N,N'-Bis(2-chlorobenzyl)ethane-1,2-diamine Dihydrochloride |
| Frakt skick | Room Temperature |
| Molekylformel | C16 H18 Cl2 N2 . 2 Cl H |
| Certifikat och efterlevnad | ISO 17025 |
| Synonym | N,N'-Bis(2-chlorobenzyl)ethane-1,2-diamine Dihydrochloride, Ticlopidine Hydrochloride Imp. J (EP) as Dihydrochloride, Ticlopidine Hydrochloride Imp. J (EP), Ticlopidine Imp. J (EP), Ticlopidine Hydrochloride Impurity J as Dihydrochloride, Ticlopidine Impurity J as Dihydrochloride |
| InChI formel | InChI=1S/C16H18Cl2N2.2ClH/c17-15-7-3-1-5-13(15)11-19-9-10-20-12-14-6-2-4-8-16(14)18;;/h1-8,19-20H,9-12H2;2*1H |
| LEDER | Cl.Cl.Clc1ccccc1CNCCNCc2ccccc2Cl |
| IUPAC-namn | N,N'-bis[(2-chlorophenyl)methyl]ethane-1,2-diamine;dihydrochloride |
| Visa mer |