Läs mer
Tiemonium Methyl Sulfate, TRC
Produktkod. 30448950
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30448950 25 mg
1 options
Produktkod. 30448950 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT43876025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Tiemonium Methyl Sulfate
CAS 6504-57-0
Molekylformel C18 H24 N O2 S . C H3 O4 S
Kvantitet 25 mg
Synonym 4-[3-Hydroxy-3-phenyl-3-(2-thienyl)propyl]-4-methylmorpholinium methyl sulfate (7CI), Morpholinium','4-[3-hydroxy-3-phenyl-3-(2-thienyl)propyl]-4-methyl-','methyl sulfate (salt) (8CI,9CI), Tiemonium methosulfate, Tiemonium methosulphate
InChI formel InChI=1S/C18H24NO2S.CH4O4S/c1-19(11-13-21-14-12-19)10-9-18(20,17-8-5-15-22-17)16-6-3-2-4-7-16;1-5-6(2,3)4/h2-8,15,20H,9-14H2,1H3;1H3,(H,2,3,4)/q+1;/p-1
LEDER COS(=O)(=O)[O-].C[N+]1(CCC(O)(c2ccccc2)c3cccs3)CCOCC1
IUPAC-namn 3-(4-methylmorpholin-4-ium-4-yl)-1-phenyl-1-thiophen-2-ylpropan-1-ol;methyl sulfate
Molekylvikt (g/mol) 429.551
Formel vikt 429.128