missing translation for 'onlineSavingsMsg'
Learn More

TIGIT, APC, clone: MBSA43, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 17-9500-42

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15508256

  • 3095.00 SEK / 100 tests


EU Stock 14
Add to basket

Description

Description

Description: The MBSA43 monoclonal antibody reacts with human T cell immunoreceptor with Ig and ITIM domains (TIGIT), which is also known as VSTM3 or WUCAM. This 26kDa protein is a member of the poliovirus receptor (PVR) family. The expression of TIGIT has been reported on NK cells, regulatory T cells, follicular T helper cells, memory CD4+ T cells, and CD8+ T cells, but it is not expressed on B cells or naive CD4+ T cells. TIGIT may be upregulated on naive CD4+ T cells upon activation. TIGIT can interact with certain members of the PVR and PVR-like families, including CD155, and also mediates the interaction of NK and T cells with antigen presenting cells, fibroblasts, and endothelial cells that express PVR and PVR-like proteins.MBSA43 is reported to crossreact with Rhesus macaque.Applications Reported: This MBSA43 antibody has been reported for use in flow cytometric analysis.Applications Tested: This MBSA43 antibody has been pre-titrated and tested by flow cytometeric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

TIGIT
Monoclonal
5μl/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
Q495A1
VSIG9, VSTM3, WUCAM, V-set and immunoglobulin domain containing 9, V-set and immunoglobulin domain-containing protein 9, V-set and transmembrane domain containing 3, V-set and transmembrane domain-containing protein 3, Washington University cell adhesion molecule
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
MBSA43
APC
TIGIT
Liquid
TIGIT
IgG1, kappa, kappa
100 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
201633
Videos

For Research Use Only.