missing translation for 'onlineSavingsMsg'
Learn More

TIGIT Mouse anti-Human, PE-Cyanine7, Clone: MBSA43, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 25-9500-41

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15568666

  • 1452.00 SEK / 25 tests


In stock, ready to ship: 9
Add to basket

Description

Description

Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 µL of cell sample + 100 µL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically. Excitation: 488-561 nm; Emission: 775 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered. Binds with high affinity to the poliovirus receptor (PVR) which causes increased secretion of IL10 and decreased secretion of IL12B and suppresses T cell activation by promoting the generation of mature immunoregulatory dendritic cells.
Specifications

Specifications

TIGIT
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
Q495A1
VSIG9, VSTM3, WUCAM, V-set and immunoglobulin domain containing 9, V-set and immunoglobulin domain-containing protein 9, V-set and transmembrane domain containing 3, V-set and transmembrane domain-containing protein 3, Washington University cell adhesion molecule
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
MBSA43
PE-Cyanine7
TIGIT
Liquid
TIGIT
IgG1, kappa
25 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
201633
Videos
SDS
Documents

For Research Use Only.