Learn More
TIGIT Monoclonal Antibody (GIGD7), Super Bright™ 702, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
1370.00 SEK - 3220.00 SEK
Specifications
Antigen | TIGIT |
---|---|
Clone | GIGD7 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15933784
|
Affymetrix eBioscience
67-9501-82 |
100 μg |
3220.00 SEK
100µg |
Estimated Shipment: 10-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15923784
|
Invitrogen
67-9501-80 |
25 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Rat Monoclonal Antibody
TIGIT Monoclonal antibody specifically detects TIGIT in Mouse samples. It is validated for Flow Cytometry.Specifications
TIGIT | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with BSA and 0.09% sodium azide; pH 7.2 | |
100043314 | |
IgG2a κ | |
Affinity chromatography | |
Antibody |
GIGD7 | |
Flow Cytometry | |
Super Bright 702 | |
Rat | |
Mouse | |
P86176 | |
TIGIT | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
TIGIT |