Läs mer
Tilbroquinol, TRC
Produktkod. 30011545
Change view
Kvantitet:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30011545 50 mg
30006218 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30011545 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT44014050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 7175-09-9
Molekylformel C10H8BrNO
Molekylvikt (g/mol) 238.08
Synonym 5-Methyl-7-bromo-8-hydroxyquinoline, 5-Methyl-7-bromooxine, 8-Hydroxy-7-bromo-5-methylquinoline, Tilbroquinol;
IUPAC-namn 7-bromo-5-methylquinolin-8-ol
LEDER Cc1cc(Br)c(O)c2ncccc12
Specifikationer
Kemiskt namn eller material Tilbroquinol
Kvantitet 50 mg
InChI formel InChI=1S/C10H8BrNO/c1-6-5-8(11)10(13)9-7(6)3-2-4-12-9/h2-5,13H,1H3
Formel vikt 236.9789