Läs mer
Tocris Bioscience™ Tilfrinib
Potent and selective breast tumor kinase (Brk) inhibitor
Brand: Tocris Bioscience™ 5579/10
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Tilfrinib is a potent and selective breast tumor kinase (Brk, also known as tyrosine-protein kinase 6, PTK6) inhibitor (IC50 = 3.15 nM). Exhibits >1000-fold selectivity for Brk over a panel of other kinases. Inhibits proliferation of breast cancer cells in vitro.Specifikationer
| 3-(9H-Pyrido[2,3-b]indol-4-ylamino)phenol | |
| 1600515-49-8 | |
| 10 mg |