Läs mer
Tilmicosin, MedChemExpress
Kvantitet:
100 mg
Beskrivning
Tilmicosin is a macrolide antibiotic, is used for the research of bovine respiratory disease and ovine respiratory disease associated with Mannheimia (Pasteurella) haemolytica.
Specifications
Specifications
| Kemiskt namn eller material | Tilmicosin |
| Fysisk form | Solid |
| CAS | 108050-54-0 |
| Färg | White |
| Kvantitet | 100 mg |
| Anteckningar om renhetsgrad | Research |
| Molekylformel | C46H80N2O13 |
| För användning med (applikation) | COVID-19-immunoregulation |
| Synonym | LY-177370 EL-870 |
| Löslighetsinformation | DMSO : 100 mg/mL (115.06 mM; Need ultrasonic) |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.