Learn More
TIM-4 Monoclonal Antibody (54 (RMT4-54)), Alexa Fluor™ 488, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
Brand: Affymetrix eBioscience 53-5866-82
Additional Details : Weight : 0.01000kg
Description
Description: The monoclonal antibody 54 recognizes mouse Tim-4 (T cell, Ig, mucin). Unlike other Tim family members, Tim-4 is not expressed on T lymphocytes but is found on antigen presenting cells such as dendritic cells and macrophages. Tim-4 has been shown to interact with Tim-1 thereby suggesting a role of Tim-4 in the balance of Th1/Th2 regulation. Additionally Tim-4 has been shown to be a receptor for phosphatidylserine and may play a role in engulfment of apoptotic cells. Expression is found on a subset of CD11c+ splenic cells. Applications Reported: This 54 (RMT4-54) antibody has been reported for use in flow cytometric analysis. Applications Tested: This 54 (RMT4-54) antibody has been tested by flow cytometric analysis of resident peritoneal macrophages. This may be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488 nm; Emission: 519 nm; Laser: Blue Laser
The T cell immunoglobulin and mucin domain containing protein (TIM) family encodes cell surface receptors that are involved in the regulation of T helper (Th) -1 and -2 cell-mediated immunity. TIM-4, which is preferentially expressed on macrophages and dendritic cells, is the natural ligand of TIM-1, and this binding leads to T-cell expansion and cytokine production. Unlike other members of the TIM family, TIM-4 lacks a putative tyrosine phosphorylation signal sequence in its intracellular domain. The TIM-4 gene maps to a locus associated with predisposition to asthma in both mice and humans and with its connection to TIM-1-triggered Th2 responsiveness, may be considered as a candidate disease/predisposition gene for asthma.Specifications
TIM-4 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
Q6U7R4 | |
TIMD4 | |
Affinity chromatography | |
RUO | |
276891 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
54 (RMT4-54) | |
Alexa Fluor 488 | |
TIMD4 | |
B430010N18Rik; RGD1564516; smuckler; soluble TIM 4; Spleen, mucin-containing, knockout of lymphotoxin protein; sTIM 4; T cell immunoglobulin and mucin domain containing 4; T-cell immunoglobulin and mucin domain containing 4; T-cell immunoglobulin and mucin domain-containing protein 4; T-cell immunoglobulin mucin receptor 4; T-cell membrane protein 4; Tim4; TIM-4; TIMD4; TIMD-4 | |
Rat | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2a κ |