Learn More
TIM-4 Monoclonal Antibody (54 (RMT4-54)), Alexa Fluor™ 488, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
1716.00 SEK - 3935.00 SEK
Specifications
Antigen | TIM-4 |
---|---|
Clone | 54 (RMT4-54) |
Concentration | 0.5 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16696404
|
Affymetrix eBioscience
53-5866-80 |
25 μg |
1716.00 SEK
Pack of 2 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16606414
|
Affymetrix eBioscience
53-5866-82 |
100 μg |
3935.00 SEK
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Description: The monoclonal antibody 54 recognizes mouse Tim-4 (T cell, Ig, mucin). Unlike other Tim family members, Tim-4 is not expressed on T lymphocytes but is found on antigen presenting cells such as dendritic cells and macrophages. Tim-4 has been shown to interact with Tim-1 thereby suggesting a role of Tim-4 in the balance of Th1/Th2 regulation. Additionally Tim-4 has been shown to be a receptor for phosphatidylserine and may play a role in engulfment of apoptotic cells. Expression is found on a subset of CD11c+ splenic cells. Applications Reported: This 54 (RMT4-54) antibody has been reported for use in flow cytometric analysis. Applications Tested: This 54 (RMT4-54) antibody has been tested by flow cytometric analysis of resident peritoneal macrophages. This may be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488 nm; Emission: 519 nm; Laser: Blue Laser
TIM-4 Monoclonal antibody specifically detects TIM-4 in Mouse samples. It is validated for Flow Cytometry.Specifications
TIM-4 | |
0.5 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
B430010N18Rik; RGD1564516; smuckler; soluble TIM 4; Spleen, mucin-containing, knockout of lymphotoxin protein; sTIM 4; T cell immunoglobulin and mucin domain containing 4; T-cell immunoglobulin and mucin domain containing 4; T-cell immunoglobulin and mucin domain-containing protein 4; T-cell immunoglobulin mucin receptor 4; T-cell membrane protein 4; Tim4; TIM-4; TIMD4; TIMD-4 | |
TIMD4 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
TIMD4 |
54 (RMT4-54) | |
Flow Cytometry | |
Alexa Fluor 488 | |
Rat | |
Mouse | |
Q6U7R4 | |
276891 | |
IgG2a κ | |
Affinity chromatography | |
Antibody |