Learn More
Abnova™ TIMM9 Recombinant Protein
Brand:  Abnova™ H00026520-P01.25ug
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16118362
  • 6845.00 SEK / 25µg
Estimated Shipment: 29-05-2024
to see stock.
Description
Description
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Specifications
Specifications
AAH20213
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
35.53
8
Glutathione Sepharose 4 Fast Flow
25 ug
MAAQIPESDQIKQFKEFLGTYNKLTETCFLDCVKDFTTREVKPEETTCSEHCLQKYLKMTQRISMRFQEYHIQQNEALAAKAGLLGQPR
TIM9/TIM9A
TIMM9
Wheat Germ (in vitro)
GST
Antibody Production, Protein Array, ELISA, Western Blot
26520
TIMM9 (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
TIMM9
Human
Recombinant
Solution