missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIMP1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16644-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
TIMP1 (metalloproteinase inhibitor 1) is a metalloproteinase inhibitor that functions by forms one-to-one complexes with target metallproteinases, such as collagenases, and irreversible inactivates them by binding to their catalytic zinc cofactor. TIMP1 acts on MMP1, MMP2, MMP3, MMP7, MMP8, MMP9, MMP10, MMP11, MMP12, MMP14, and MMP16. It also functions as a growth factor that regulates cell differenation, migration, and cell death. TIMP1 activates cellular signaling cascades via CD63 and ITGB1.Specifications
TIMP1 | |
Polyclonal | |
Unconjugated | |
TIMP1 | |
CLGI, Collagenase inhibitor, EPA, EPO, HCI, Metalloproteinase inhibitor 1, TIMP, TIMP 1, TIMP1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
7076 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P01033 | |
TIMP1 | |
TIMP1 Fusion Protein Ag10006 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |