Learn More
Tin AAS Standard, 1000 mg/L Sn in HCl 20%, Honeywell Fluka™
Brand: Honeywell Fluka E4071-500ML
Additional Details : Weight : 0.55500kg Transport : UN number : 1789 Chem class : 8 Pack group : III
Description
Honeywell Fluka™ analytical standards offer a growing range of high quality inorganic and organic reference materials for the European market produced according to ISO 9001. The products are suitable for a wide range of industries and applications and come complete with a detailed Certificate of Analysis (CoA) in every delivery, and are eligible for free expert technical support ensuring you have the information you need to quickly start your analysis and pass an audit.
Specifications
Tin AAS Standard | |
1.11 g/cm3 | |
500 mL |
Standards and Modifiers for AAS | |
<2 |