Läs mer
Tinopal RBS 200, TRC
Produktkod. 30455855
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30455855 50 mg
1 options
Produktkod. 30455855 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT44409550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Tinopal RBS 200
CAS 6416-68-8
Molekylformel C24 H16 N3 Na O3 S
Kvantitet 50 mg
Synonym 4-(2H-Naphtho[1,2-d]triazol-2-yl)-2-stilbenesulfonic Acid Sodium Salt, ,'5-(2H-Naphtho[1,2-d]triazol-2-yl)-2-(2-phenylethenyl)-benzenesulfonic Acid Sodium Salt, C.I. Fluorescent Brightening Agent 46','Sodium Salt, 2H-Naphtho[1,2-d]triazole','benzenesulfonic acid deriv., 2-(4-Styryl-3-sulfophenyl)-2H-naphtho[1,2-d]triazole Sodium Salt, 4-(Naphtho-1',2', 4,5-triazol-2-yl)-2-sulfostilbene Sodium Salt, C.I. 40645, C.I. Fluorescent Brightener 46, Fluorescent Brightener 46, Pylaklor White S 15A, Sodium 2-(4-styryl-3-sulfophenyl)-2H-naphtho[1,2-d]triazole, Sodium 4-(2H-Naphtho[1,2-d]triazol-2-yl)stilbene-2-sulfonate, Tinopal RBS
InChI formel InChI=1S/C24H17N3O3S.Na/c28-31(29,30)23-16-20(14-12-19(23)11-10-17-6-2-1-3-7-17)27-25-22-15-13-18-8-4-5-9-21(18)24(22)26-27;/h1-16H,(H,28,29,30);/q;+1/p-1/b11-10+;
LEDER [Na]OS(=O)(=O)c1cc(ccc1\C=C\c2ccccc2)n3nc4ccc5ccccc5c4n3
IUPAC-namn [5-benzo[e]benzotriazol-2-yl-2-[(E)-styryl]phenyl]sulfonyloxysodium
Molekylvikt (g/mol) 449.46
Formel vikt 449.081