Learn More
Lovibond™ Tintometer™ Ammonium Molybdate Solution, 100mL
For use with Lovibond test kits, colorimeters and spectrophotometers
Brand: Lovibond™ 460241
Additional Details : Weight : 0.30000kg
Description
- Ammonium Molybdate Solution
- 100mL
Specifications
Ammonia Test | |
Test kits, colorimeters and spectrophotometers | |
100 mL | |
Ammonium Molybdate Solution |
Safety and Handling
- 100ML AMMONIA MOLYBDATE SOLUTION .
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 2
Hazard Statement
- H290-May be corrosive to metals.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P390-Absorb spillage to prevent material damage.
Supplemental information
- MIXTURE LIST-Contains : sulphuroc acid