Learn More
Lovibond™ Tintometer™ CHECKIT™ Comparator, Copper Vario HR Test, 0 to 0.5mg/L
Suitable for both mobile and static water analysis. Checkit™ Comparators are available with a number of different color scales and provide the basis for a comprehensive easy to to to use analysis system.
Brand:  Lovibond™ 147431
Additional Details : Weight : 0.74000kg
Product Code. 10359934
  • 1255.00 SEK / Each
Estimated Shipment: 29-05-2024
to see stock.
Add to basket
Description
Description
  • The Lovibond™ Checkit™ comparator is a compact, lightweight colorimetric unit
  • Suitable for both mobile and static water analysis
  • Available with a number of different color scales
  • Provides the basis for a comprehensive easy to to to use analysis system
  • Each Checkit™ Comparator Disc contains a continuous colour scale for exact color matching between standard and sample
  • Each Checkit™ Comparator Kit is supplied in a carry case with comparator, vials, discs, reagents and method instructions for all tests
  • The Copper disc supplied measures from 0 to 5mg/L Free Copper with the following steps: 0 / 0.5 / 1.0 / 1.5 / 2.0 / 2.5 / 3.0 / 3.5 / 4.0 / 5.0
Specifications
Specifications
Water Quality Testing
Carry case with Comparator, Vials, Discs, reagents and method instructions for all tests
Copper Vario HR Test
Checkit™ Comparator
0 to 5mg/L