Learn More
Lovibond™ Tintometer™ Comparator 2000+ Disc 3/29A, Sugar Test, 1 to 10ppm
Glass color standards are fade-free, resistant to chemicals and scratchproof. Use these discs with the Comparator 2000+ for color comparator tests.
Brand: Lovibond™ 232910
Additional Details : Weight : 0.09000kg
Description
- The required accuracy of results is only ensured if stable, fade-free color standards are used
- Glass color standards are fade-free, resistant to chemicals and scratchproof
- Lovibond™ standards are made from colored glass filters
- Comply with international standards, e.g. ISO 7393/2
- Readings: 0, 5, 10, 15, 30, 45, 60, 75, 100mg/L
Specifications
1 to 10ppm | |
Comparator 2000+ | |
Sugar Test | |
Comparator 2000+ Disc 3/29A |