Learn More
Lovibond™ Tintometer™ Potassium T Tablets, 0.7 to 12mg/L
For use with Lovibond test kits, colorimeters and spectrophotometers. Using tablet reagents guarantees exact reagent dosing, allowing for more precise titrations than traditional drop count test kits
Brand: Lovibond™ 515670BT
Additional Details : Weight : 0.50000kg
Description
- Range: 0.7 to 12mg/L
- Parameter: Potassium
- Method: Tetraphenylborate-turbidity
- Case of 100
Specifications
Photometry Tablet Reagents | |
Test kits, colorimeters and spectrophotometers | |
100 Tablets | |
Potassium T Tablets |