Läs mer
Tocris Bioscience™ Tiotropium bromide
Potent muscarinic receptor antagonist
Brand: Tocris Bioscience™ 5902/50
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Tiotropium bromide is a potent muscarinic receptor antagonist. Inhibits electrical field stimulation-induced contraction of guinea pig trachea in vitro (IC50 = 0.17 nM). Induces smooth muscle relaxation and bronchodilation.Specifikationer
| (1α,2β,4β,5α,7β)-7-[(2-Hydroxy-2,2-di-2-thienylacetyl)oxy]-9,9-dimethyl-3-oxa-9-azoniatricyclo[3.3.1.02,4]nonane bromide | |
| 136310-93-5 | |
| Ba 679 BR | |
| 50 mg |