Läs mer
Tocris Bioscience™ Tiotropium bromide
Beskrivning
Tiotropium bromide is a potent muscarinic receptor antagonist. Inhibits electrical field stimulation-induced contraction of guinea pig trachea in vitro (IC50 = 0.17 nM). Induces smooth muscle relaxation and bronchodilation.
Specifikationer
Specifikationer
| Kemiskt namn eller material | (1α,2β,4β,5α,7β)-7-[(2-Hydroxy-2,2-di-2-thienylacetyl)oxy]-9,9-dimethyl-3-oxa-9-azoniatricyclo[3.3.1.02,4]nonane bromide |
| CAS | 136310-93-5 |
| Synonym | Ba 679 BR |
| Kvantitet | 50 mg |
| Mål | Non-selective mAChR Antagonists |
| Molekylformel | C19H22BrNO4S2 |
| Formel vikt | 472.42 |
| Renhet | >98% |