Läs mer
Tiotropium Bromide, TRC
Produktkod. 30135309
Change view
Kvantitet:
1 mg
5 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30135309 100 mg
30004234 1 mg
30010561 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30135309 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT444850100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 136310-93-5
Molekylformel C19 H22 N O4 S2 . Br
Molekylvikt (g/mol) 472.416
Synonym 3-Oxa-9-azoniatricyclo[3.3.1.02,4]nonane, 7-[(2-hydroxy-2,2-di-2-thienylacetyl)oxy]-9,9-dimethyl-, bromide (1:1), (1α,2β,4β,5α,7β)-, 3-Oxa-9-azoniatricyclo[3.3.1.02,4]nonane, 7-[(hydroxydi-2-thienylacetyl)oxy]-9,9-dimethyl-, bromide, (1α,2β,4β,5α,7β)- (9CI), 3-Oxa-9-azatricyclo[3.3.1.02,4]nonane, 3-oxa-9-azoniatricyclo[3.3.1.02,4]nonane deriv., (1α,2β,4β,5α,7β)-7-[(Hydroxydi-2-thienylacetyl)oxy]-9,9-di-methyl-3-oxa-9-azoniatricyclo[3.3.1.0]nonane bromide, BA 679BR, Spiriva, Tiotropium bromide, tiopropium
LEDER [Br-].C[N+]1(C)[C@@H]2C[C@H](C[C@H]1[C@@H]3O[C@H]23)OC(=O)C(O)(c4cccs4)c5cccs5
Specifikationer
Kemiskt namn eller material Tiotropium Bromide
Kvantitet 100 mg
InChI formel InChI=1S/C19H22NO4S2.BrH/c1-20(2)12-9-11(10-13(20)17-16(12)24-17)23-18(21)19(22,14-5-3-7-25-14)15-6-4-8-26-15;/h3-8,11-13,16-17,22H,9-10H2,1-2H3;1H/q+1;/p-1/t11-,12-,13+,16-,17+;
Formel vikt 471.017