Läs mer
Tiotropium EP Impurity B, Mikromol™
Produktkod. 30052448
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30052448 25 mg
30049070 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30052448 Leverantör Mikromol Leverantörsnummer MM1293.020025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Tiotropium EP Impurity B
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (1R,2R,4S,5S,7s)-9-Methyl-3-oxa-9-azatricyclo(3.3.1.02,4)nonan-7-yl 2-Hydroxy-2,2-dithiophen-2-ylacetate
CAS 136310-64-0
Analyt- eller komponentnamn (1R,2R,4S,5S,7s)-9-Methyl-3-oxa-9-azatricyclo(3.3.1.02,4)nonan-7-yl 2-Hydroxy-2,2-dithiophen-2-ylacetate
Frakt skick Room Temperature
Molekylformel C18 H19 N O4 S2
Certifikat och efterlevnad ISO 9001
Synonym (1R,2R,4S,5S,7s)-9-Methyl-3-oxa-9-azatricyclo[3.3.1.02,4]nonan-7-yl 2-Hydroxy-2,2-dithiophen-2-ylacetate, 2-Thiopheneacetic acid, alpha-hydroxy-alpha-2-thienyl-, 9-methyl-3-oxa-9-azatricyclo[3.3.1.02,4]non-7-yl ester, (1alpha,2beta,4beta,5alpha,7beta)-, (1alpha,2beta,4beta,5alpha,7beta)-7-[(Hydroxydi(2-thienyl)acetyl)oxy]-9-methyl-3-oxa-9-azatricyclo[3.3.1.0]nonane, Di(2-thienyl)glycolic acid scopine ester, Dithienylglycolic acid scopine ester, N-Demethyl tiotropium, Scopine di(2-thienylglycolate), Scopine hydroxydi(2-thienyl)acetate, 2-Thiopheneacetic acid, alpha-hydroxy-alpha-2-thienyl-, (1alpha,2beta,4beta,5alpha,7beta)-9-methyl-3-oxa-9-azatricyclo[3.3.1.02,4]non-7-yl ester, Tiotropium Bromide Monohydrate Imp B (EP), Tiotropium Bromide Monohydrate Impurity B, Tiotropium Impurity B
InChI formel InChI=1S/C18H19NO4S2/c1-19-11-8-10(9-12(19)16-15(11)23-16)22-17(20)18(21,13-4-2-6-24-13)14-5-3-7-25-14/h2-7,10-12,15-16,21H,8-9H2,1H3/t10-,11-,12+,15-,16+
LEDER CN1[C@@H]2C[C@H](C[C@H]1[C@@H]3O[C@H]23)OC(=O)C(O)(c4cccs4)c5cccs5
IUPAC-namn (1R,2R,4S,5S,7s)-9-Methyl-3-oxa-9-azatricyclo[3.3.1.02,4]nonan-7-yl 2-Hydroxy-2,2-dithiophen-2-ylacetate
Molekylvikt (g/mol) 377.48
Kvantitet 25 mg
Formel vikt 377.0755 g/mol