Läs mer
Tipiracil Hydrochloride, TRC
Produktkod. 30454188
Change view
Kvantitet:
5 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30454188 5 mg
30452737 25 mg
30449806 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30454188 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT4448905MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 183204-72-0
Molekylformel C9H11ClN4O2 . (HCl)
Molekylvikt (g/mol) 279.12
Synonym 5-Chloro-6-[(2-imino-1-pyrrolidinyl)methyl]-2,4(1H,3H)-pyrimidinedione Monohydrochloride, 2,4(1H,3H)-Pyrimidinedione, 5-chloro-6-[(2-imino-1-pyrrolidinyl)methyl]-, monohydrochloride (9CI), 5-Chloro-6-[(2-iminopyrrolidin-1-yl)methyl]pyrimidine-2,4(1H,3H)-dione hydrochloride, TAS 1-462, TPI hydrochloride, 2,4(1H,3H)-Pyrimidinedione, 5-chloro-6-[(2-imino-1-pyrrolidinyl)methyl]-, hydrochloride (1:1)
IUPAC-namn 5-chloro-6-[(2-iminopyrrolidin-1-yl)methyl]-1H-pyrimidine-2,4-dione;hydrochloride
LEDER Cl.ClC1=C(CN2CCCC2=N)NC(=O)NC1=O
Specifikationer
Kemiskt namn eller material Tipiracil Hydrochloride
Kvantitet 5 mg
InChI formel InChI=1S/C9H11ClN4O2.ClH/c10-7-5(12-9(16)13-8(7)15)4-14-3-1-2-6(14)11;/h11H,1-4H2,(H2,12,13,15,16);1H
Formel vikt 278.0337