Learn More
Kartell™ Tissue Embedding Cassettes - Uni Type
For paraffin inclusions, Universal type, provided with clipon closure lid that can easily be removed
Brand:  Kartell™ 0292104
Additional Details : Weight : 6.77000kg
Product Code. 15927860
  • 2275.00 SEK / Pack of 2000
Estimated Shipment: 27-06-2024
to see stock.
Description
Description
  • CE marked, IVD ceritfied
  • Universal Type
  • Clipon closure lid that can easily be removed
Specifications
Specifications
2000/Pk.
40 mm
Polyoxymethylene (POM)
Blue
28 mm