Läs mer
SI Analytics™ Titration Stand With Pump
Produktkod. 10369860 Handla allt SI Analytics Produkter
Change view
Spänning:
110 till 240V
Förpackningsstorlek:
Varje
Produktkod. Spänning unitSize
10369860 110 till 240V Varje
1 options
Produktkod. 10369860 Leverantör SI Analytics™ Leverantörsnummer 285216611
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
SI Analytics™ Titration Stand With Pump includes a reagent bottle, a waste bottle, and a moisture bottle.
  • Titration stand with pump
  • Basic unit with 1 l DURAN ™-reagent bottle TZ 1791, 
  • 1 l DURAN™-waste bottle TZ 1792, 
  • moisture bottle, tubes and screw threads, power supply TZ 1855 (110 ... 240 V) 
TRUSTED_SUSTAINABILITY
Specifikationer
Typ Titration Stand With Pump
Inkluderar 1 DURAN™ Reagent Bottle TZ 1791, 1 DURAN™ Waste bottle TZ 1792, Moisture Bottle, Tubes and Screw Threads, Power Supply TZ 1855 (110 to 240V)
Spänning 110 till 240V