Läs mer
Tixocortol 21-Pivalate, TRC
Produktkod. 30134206
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134206 100 mg
30008642 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30134206 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT447500100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 55560-96-8
Molekylformel C26 H38 O5 S
Molekylvikt (g/mol) 462.64
Synonym Pregn-4-ene-3,20-dione, 21-[(2,2-dimethyl-1-oxopropyl)thio]-11,17-dihydroxy-, (11β)-, (11β)-21-[(2,2-Dimethyl-1-oxopropyl)thio]-11,17-dihydroxypregn-4-ene-3,20-dione, JO 1016, Pivalone, Rectovalone, Tiovalon, Tixocortol 21-pivalate, Tixocortol pivalate
IUPAC-namn S-[2-[(8S,9S,10R,11S,13S,14S,17R)-11,17-dihydroxy-10,13-dimethyl-3-oxo-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-17-yl]-2-oxoethyl] 2,2-dimethylpropanethioate
LEDER CC(C)(C)C(=O)SCC(=O)[C@@]1(O)CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3[C@@H](O)C[C@]12C
Specifikationer
Kemiskt namn eller material Tixocortol 21-Pivalate
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C26H38O5S/c1-23(2,3)22(30)32-14-20(29)26(31)11-9-18-17-7-6-15-12-16(27)8-10-24(15,4)21(17)19(28)13-25(18,26)5/h12,17-19,21,28,31H,6-11,13-14H2,1-5H3/t17-,18-,19-,21+,24-,25-,26-/m0/s1
Formel vikt 462.244
Procent renhet >95%