Läs mer
Tixocortol, TRC
Produktkod. 30136502
Change view
Kvantitet:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136502 10 mg
30012294 50 mg
30008978 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30136502 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT44749010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 61951-99-3
Molekylformel C21 H30 O4 S
Molekylvikt (g/mol) 378.53
Synonym Pregn-4-ene-3,20-dione, 11,17-dihydroxy-21-mercapto-, (11beta)-, Progesterone, 11beta,17-dihydroxy-21-mercapto- (6CI), (11beta)-11,17-Dihydroxy-21-mercaptopregn-4-ene-3,20-dione, Cortisol 21-thiol, Tixocortol
IUPAC-namn (8S,9S,10R,11S,13S,14S,17R)-11,17-dihydroxy-10,13-dimethyl-17-(2-sulfanylacetyl)-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-3-one
LEDER C[C@]12CCC(=O)C=C1CC[C@H]3[C@@H]4CC[C@](O)(C(=O)CS)[C@@]4(C)C[C@H](O)[C@H]23
Specifikationer
Kemiskt namn eller material Tixocortol
Kvantitet 10 mg
InChI formel InChI=1S/C21H30O4S/c1-19-7-5-13(22)9-12(19)3-4-14-15-6-8-21(25,17(24)11-26)20(15,2)10-16(23)18(14)19/h9,14-16,18,23,25-26H,3-8,10-11H2,1-2H3/t14-,15-,16-,18+,19-,20-,21-/m0/s1
Formel vikt 378.1865